Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-oxo-L-proline methyl ester, 97%

Catalog No. AC449272500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449272500 250 mg Glass bottle
AC449270010 1 g Glass bottle
AC449270050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449272500 Supplier Thermo Scientific Chemicals Supplier No. 449272500

CAS: 102195-80-2 | C11H17NO5 | 243.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102195-80-2
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.26
InChI Key UPBHYYJZVWZCOZ-QMMMGPOBSA-N
Synonym boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
PubChem CID 10988485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC

Specifications

Melting Point 42°C to 45°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 243.26
Percent Purity 97%
Chemical Name or Material N-BOC-4-oxo-L-proline methyl ester
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.