Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-alanine methyl ester, 98%, Thermo Scientific™

Catalog No. AAH6174203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6174203 1 g
AAH6174206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6174203 Supplier Thermo Scientific Chemicals Supplier No. H6174203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91103-47-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00191865
InChI Key GJDICGOCZGRDFM-ZCFIWIBFSA-N
Synonym boc-d-ala-ome, boc-d-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino propanoate, n-boc-d-alanine methyl ester, n-tert-butoxycarbonyl-d-alanine methyl ester, d-alanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino propanoate, boc-d-ala.ome, n-t-boc-d-alanine methyl ester
PubChem CID 637612
IUPAC Name methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C(=O)OC)NC(=O)OC(C)(C)C

Specifications

Melting Point 34°C to 37°C
Color White
Beilstein 4310313
Density 1.030 g/mL
Quantity 1 g
Flash Point 113°C (235°F)
Formula Weight 203.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-D-alanine methyl ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.