Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-aspartic acid 1-tert-butyl ester, 98%

Catalog No. AAH66716MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66716MD 250 mg
AAH6671603 1 g
AAH6671606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66716MD Supplier Thermo Scientific Chemicals Supplier No. H66716MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 77004-75-2 | C13H23NO6 | 289.328 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77004-75-2
Molecular Formula C13H23NO6
Molecular Weight (g/mol) 289.328
MDL Number MFCD00236827
InChI Key RAUQRYTYJIYLTF-MRVPVSSYSA-N
Synonym boc-d-asp-otbu, r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-boc-d-aspartic acid 1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoic acid, 3r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-d-aspotbu, n-boc-d-asparticacid1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoicacid, n-tert-butyloxycarbonyl-d-aspartic acid 1-tert-butyl ester
PubChem CID 7019660
IUPAC Name (3R)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C

Specifications

Beilstein 3553120
Quantity 250 mg
Formula Weight 289.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-aspartic acid 1-tert-butyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.