Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6288906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6288906 5 g
AAH6288914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6288906 Supplier Thermo Scientific Chemicals Supplier No. H6288906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 31972-52-8 | Molecular Formula: C9H16N2O5 | Molecular Weight: 232.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31972-52-8
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038558
InChI Key HWBAHOVOSOAFLE-UHFFFAOYSA-N
Synonym boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly
PubChem CID 333466
IUPAC Name 2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)NCC(=O)NCC(=O)O

Specifications

Color White
Linear Formula (CH3)3COCONHCH2CONHCH2CO2H
Quantity 5 g
Formula Weight 232.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-glycylglycine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.