Learn More
N-Boc-guanidine, 95%
Description
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.
Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 170°C to 175°C |
| Color | White |
| Linear Formula | (CH3)3OCONHC(=NH)NH2 |
| Quantity | 1 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in most alcohols,esters (e.g.,ethyl acetate),ketones (e.g.,acetone),acetonitrile,and dichloromethane. Not soluble in diethyl ether and hexane. |
| Formula Weight | 159.19 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | N-Boc-guanidine |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.