Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-glutamic acid 5-benzyl ester, 98%

Catalog No. AAH5687906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5687906 5 g
AAH5687914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5687906 Supplier Thermo Scientific Chemicals Supplier No. H5687906
Add to Cart

CAS: 13574-13-5 | C17H22NO6 | 336.37 g/mol

N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

Solubility
Soluble in N,N- Dimethyl formamide.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13574-13-5
Molecular Formula C17H22NO6
Molecular Weight (g/mol) 336.37
MDL Number MFCD00065569
InChI Key AJDUMMXHVCMISJ-ZDUSSCGKSA-M
Synonym boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
PubChem CID 83589
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 69°C to 71°C
Quantity 5 g
Beilstein 2226572
Solubility Information Soluble in N,N- Dimethyl formamide.
Optical Rotation −5.5° (c=1 in Acetic Acid)
Formula Weight 337.37
Percent Purity 98%
Chemical Name or Material N-Boc-L-glutamic acid 5-benzyl ester
EINECSNumber 237-007-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.