Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-glutamic acid 5-tert-butyl ester, 99%

Catalog No. AAB2232203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2232203 1 g
AAB2232206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2232203 Supplier Thermo Scientific Chemicals Supplier No. B2232203
Add to Cart

CAS: 13726-84-6 | C14H25NO6 | 303.355 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-84-6
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.355
MDL Number MFCD00038257
InChI Key YGSRAYJBEREVRB-VIFPVBQESA-N
Synonym boc-glu otbu-oh, boc-l-glu otbu-oh, s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-glutamic acid 5-tert-butyl ester, n-boc-l-glutamic acid 5-tert-butyl ester, n-tert-butoxycarbonyl-l-glutamic acid gamma-tert-butyl ester, 2s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-glu otbu, ambotzbaa1297, pubchem18923
PubChem CID 6993432
IUPAC Name (2S)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Melting Point 110°C to 112°C
Quantity 1 g
Formula Weight 303.36
Percent Purity 99%
Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.