Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-L-Valinol, 96%, Thermo Scientific™

Catalog No. p-3770760
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399230010 1 g Glass bottle
AC399230050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399230010 Supplier Thermo Scientific Chemicals Supplier No. 399230010
Add to Cart

Chemical Identifiers

CAS 79069-14-0
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD00082635
InChI Key OOQRRYDVICNJGC-MRVPVSSYSA-N
Synonym n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
PubChem CID 7021464
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
SMILES CC(C)[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Density 0.9950g/mL
Boiling Point 218.0°C
Quantity 1 g
Flash Point 85°C
Assay Percent Range 96%
Formula Weight 203.28
Percent Purity 96%
Chemical Name or Material N-BOC-L-Valinol
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.