Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-O-tert-butyl-L-tyrosine, 98%

Catalog No. AAH6280106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6280106 5 g
AAH6280114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6280106 Supplier Thermo Scientific Chemicals Supplier No. H6280106
  • $179.00
    $152.65 / Each of 1

    Save $26.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 47375-34-8 | C18H27NO5 | 337.42 g/mol

N-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.

Solubility
Soluble in DMF. Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47375-34-8
Molecular Formula C18H27NO5
Molecular Weight (g/mol) 337.42
MDL Number MFCD00065598
InChI Key ZEQLLMOXFVKKCN-YQTOOIBONA-N
Synonym boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
PubChem CID 7017901
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O

Specifications

Melting Point 113°C to 118°C
Quantity 5 g
Beilstein 4327336
Solubility Information Soluble in DMF. Insoluble in water.
Optical Rotation −15° (c=1 in DMF)
Formula Weight 337.41
Percent Purity 98%
Chemical Name or Material N-Boc-O-tert-butyl-L-tyrosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.