Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%

Catalog No. AAH62495MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62495MD 250 mg
AAH6249503 1 g
AAH6249506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62495MD Supplier Thermo Scientific Chemicals Supplier No. H62495MD

CAS: 89813-47-8 | C17H23NO5 | 321.373 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89813-47-8
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD00076979
InChI Key BEIPCYKSYYZEJH-KGLIPLIRSA-N
Synonym boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
PubChem CID 11902907
IUPAC Name 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O

Specifications

Color Brown
Quantity 250 mg
Formula Weight 321.37
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline benzyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.