Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Bromosuccinimide, 98%, Spectrum™ Chemical
SDP

Catalog No. 18604763 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-763 25 kg Amber Glass Bottle
18-604-760 100 g Amber Glass Bottle
18-604-761 500 g Amber Glass Bottle
18-604-762 2.5 kg Amber Glass Bottle
18-604-764 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-763 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B115825KGBL
Add to Cart
Add to Cart
Request a Quote

B1158, 128-08-5, C4H4BrNO2

Spectrum™ Chemical N-Bromosuccinimide , also known as NBS, is a chemical reagent used in organic chemistry in electrophilic addition reactions and radical substitution. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.99
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES BrN1C(=O)CCC1=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material N-Bromosuccinimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.