Learn More
Description
Specifications
Specifications
| Melting Point | -89°C |
| Color | Colorless |
| Acidity | 0.005% max. |
| Density | 0.810g/cm³ |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | ≥99.8 % |
| Linear Formula | CH3CH2CH2CH2OH |
| Vapor Pressure | 6.7mbar at 20°C |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
| Recommended Storage | RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.