Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Acetate ASTM D4615, Reagents

Catalog No. 01337379
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-379 500 mL Glass Bottle
01-337-381 4 L Poly Bottle
01-337-380 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-379 Supplier Reagents Holdings Llc Supplier No. C2156300500C
Add to Cart
Add to Cart

n-Butyl Acetate ASTM D4615, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym Butyl Ethanoate, Acetic Acid Butyl Ester
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Boiling Range 126°C
Melting Point -78°C
Color Colorless
Boiling Point 126°C
Melting Range -78°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 0.88
Physical Form Liquid
Chemical Name or Material n-Butyl Acetate ASTM D4615
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.