Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Alcohol, for HPLC, GC and spectrophotometry, ≥99.9%, Solstice

Catalog No. 6000091 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-000-91 4 x 4 L Glass Bottle
60-000-92 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-000-91 Supplier Solstice Supplier No. 0244
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Absorbance 0.5 AU max. at 225 nm (UV), 0.04 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.81 g/cm3
Quantity 4 x 4 L
Flash Point 37°C
Packaging Glass Bottle
UN Number UN1120
Water 0.03% max.
Beilstein 969148
Boiling Point 117°C
Solubility Information 781% in Water at 20°C
Formula Weight 74.12
Percent Purity >99%
Grade HPLC
Chemical Name or Material n-Butyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butanols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.