Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Alcohol, Purified (1-Butanol), Reagents

Catalog No. 01337550
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-550 20 L
01-337-551 1 L
01-337-549 4 L
01-337-548 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-550 Supplier Reagents Holdings Llc Supplier No. C212000020E6
Add to Cart
Add to Cart

n-Butyl Alcohol, Purified (1-Butanol), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-Butanol, n-Butyl Alcohol, 1-Butyl Alcohol, Butan-1-ol
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point <88.6°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.81
Boiling Point 118°C
Grade Pure
Chemical Name or Material n-Butyl Alcohol, Purified (1-Butanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.