Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl L-lactate, 97+%

Catalog No. AAA1382130
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1382130 250 g
AAA138210B 1000 g
AAA138210I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1382130 Supplier Thermo Scientific Chemicals Supplier No. A1382130
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 34451-19-9 | C7H14O3 | 146.19 g/mol

Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.

Solubility
Soluble in alcohol and water(42 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34451-19-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD00004519,MFCD00066454
InChI Key MRABAEUHTLLEML-UHFFFAOYNA-N
PubChem CID 6999977
IUPAC Name butyl (2S)-2-hydroxypropanoate
SMILES CCCCOC(=O)C(C)O

Specifications

Density 0.984
Boiling Point 185°C to 187°C
Flash Point 69°C (157°F)
Refractive Index 1.421
Quantity 250 g
Synonym butyl 2s-2-hydroxypropanoate, unii-33glz553p8, butyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, butyl ester, 2s, butyl l-lactate, --butyl l-lactate, propanoic acid,2-hydroxy-, butyl ester, 2s, +-butyl lactate, butyl-l-lactate, butyl lactate, s
Solubility Information Soluble in alcohol and water(42g/L (25°C).
Formula Weight 146.19
Percent Purity ≥97%
Chemical Name or Material n-Butyl L-lactate
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 252-036-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.