Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl methyl sulfide, 98+%, Thermo Scientific™

Catalog No. AAL1203906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1203906 5 g
AAL1203914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1203906 Supplier Thermo Scientific Chemicals Supplier No. L1203906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-29-5
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00015256
InChI Key WCXXISMIJBRDQK-UHFFFAOYSA-N
Synonym butyl methyl sulfide, butane, 1-methylthio, n-butyl methyl sulfide, butyl methyl sulphide, sulfide, butyl methyl, 1-methylthio butane, unii-2n5imb9xap, methyl butyl sulfide, 2-thiahexane, 2n5imb9xap
PubChem CID 12339
IUPAC Name 1-methylsulfanylbutane
SMILES CCCCSC

Specifications

Density 0.85
Boiling Point 122°C to 123°C
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.4621
Linear Formula CH3S(CH2)3CH3
Quantity 5 g
UN Number UN1993
Beilstein 1730959
Formula Weight 104.22
Percent Purity ≥98%
Chemical Name or Material n-Butyl methyl sulfide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-034-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.