Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Butylbenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. B071625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B071625G 25 g
B0716500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B071625G Supplier TCI America Supplier No. B071625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3622-84-2
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD00025024
InChI Key IPRJXAGUEGOFGG-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-butyl, plastomoll bmb, n-n-butylbenzenesulfonamide, dellatol bbs, n-butyl-benzenesulfonamide, benzenesulfonic acid butyl amide, plasthall bsa, cetamoll bmb, uniplex 214, bm 4 sulfonamide
PubChem CID 19241
ChEBI CHEBI:44237
IUPAC Name N-butylbenzenesulfonamide
SMILES CCCCNS(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 184°C
Quantity 25 g
Formula Weight 213.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material N-Butylbenzenesulfonamide
EINECSNumber (3)-1894&(3)-1917&(3)-1930
RTECSNumber DB1283000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.