Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Butylbenzylamine 97.0+%, TCI America™
SDP

Catalog No. B204325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B204325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B204325ML Supplier TCI America Supplier No. B204325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2403-22-7
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00009427
InChI Key HIPXPABRMMYVQD-UHFFFAOYSA-N
Synonym n-butylbenzylamine, n-benzylbutylamine, n-benzyl-n-butylamine, benzenemethanamine, n-butyl, butylbenzylamine, benzylbutylamine, benzylamine, n-butyl, benzyl butyl amine, n-n-butyl benzylamine, benzylbutylamin
PubChem CID 75467
IUPAC Name N-benzylbutan-1-amine
SMILES CCCCNCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 2735
Formula Weight 163.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Butylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.