Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-4-aminobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. C07531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07531G 1 g
C075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07531G Supplier TCI America Supplier No. C07531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5105-78-2
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.26
MDL Number MFCD00055835
InChI Key STQMDRQJSNKUAW-UHFFFAOYSA-N
Synonym z-gamma-abu-oh, 4-benzyloxy carbonyl amino butanoic acid, n-cbz-gaba, carbobenzoxy-4-aminobutyric acid, n-cbz-4-aminobutanoic acid, z-gaba-oh, n-cbz-4-aminobutyric acid, 4-carboxyamino butyric acid n-benzyl ester, z-l-gamma-aminobutyric acid
PubChem CID 21184
IUPAC Name 4-{[(benzyloxy)carbonyl]amino}butanoic acid
SMILES OC(=O)CCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 64°C
Color White
Quantity 1 g
Formula Weight 237.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-4-aminobutyric Acid
RTECSNumber ES8137200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.