Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-alanine 98.0+%, TCI America™
SDP

Catalog No. C066425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C066425G 25 g
C06641G 1 g
C06645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C066425G Supplier TCI America Supplier No. C066425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26607-51-2
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00063126
InChI Key TYRGLVWXHJRKMT-UHFFFAOYNA-N
Synonym n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
PubChem CID 70832
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}propanoic acid
SMILES CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85°C
Color White
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.