Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-leucine Dicyclohexylammonium Salt 98.0+%, TCI America™
SDP

Catalog No. C21351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21351G 1 g
C21355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21351G Supplier TCI America Supplier No. C21351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7662-58-0
Molecular Formula C26H42N2O4
Molecular Weight (g/mol) 446.63
MDL Number MFCD10566885
InChI Key FOULZFSGIVQTHX-HVPILOLGNA-N
Synonym z-d-leu-oh.dcha, n-carbobenzoxy-d-leucine dicyclohexylammonium salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-methylpentanoate, 2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid; dicha, z-d-leu-ohcha, n-z-leu-oh.dcha, n-cbz-d-leucine dicyclohexylammonium salt, n-benzyloxycarbonyl-d-leucine.dicyclohexylamine, n-carbobenzoxy-d-leucinedicyclohexylammoniumsalt, n-benzyloxycarbonyl-d-leucine dicyclohexylammonium salt
PubChem CID 44629899
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C
Color White
Quantity 1 g
Formula Weight 446.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-leucine Dicyclohexylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.