Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-proline 98.0+%, TCI America™
SDP

Catalog No. C17305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17305G 5 g
C173025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17305G Supplier TCI America Supplier No. C17305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6404-31-5
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD00063228
InChI Key JXGVXCZADZNAMJ-LLVKDONJSA-N
Synonym n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid
PubChem CID 111208
IUPAC Name (2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 76°C
Quantity 5 g
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-proline
EINECSNumber (5)-6709
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.