Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-threonine 98.0+%, TCI America™
SDP

Catalog No. C21381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21381G 1 g
C21385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21381G Supplier TCI America Supplier No. C21381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80384-27-6
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00037808
InChI Key IPJUIRDNBFZGQN-WCBMZHEXSA-N
Synonym z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
PubChem CID 853484
IUPAC Name (2R,3S)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 1 g
Formula Weight 253.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.