Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-leucine 98.0+%, TCI America™
SDP

Catalog No. C06421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06421G 1 g
C06425G 5 g
C064225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C06421G Supplier TCI America Supplier No. C06421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3588-60-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00065129
InChI Key USPFMEKVPDBMCG-UHFFFAOYSA-N
Synonym N-Cbz-DL-leucine, Z-DL-Leu-OH
PubChem CID 246673
IUPAC Name 4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 265.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.