Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-glutamine 98.0+%, TCI America™
SDP

Catalog No. C057425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057425G 25 g
C05745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C057425G Supplier TCI America Supplier No. C057425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2650-64-8
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00008043
InChI Key JIMLDJNLXLMGLX-JTQLQIEISA-N
Synonym z-gln-oh, n-carbobenzyloxy-l-glutamine, cbz-l-glutamine, cbz-gln-oh, n-carbobenzoxy-l-glutamine, benzyloxycarbonyl-l-glutamine, carbobenzoxyglutamine, n-cbz-l-glutamine, cbz-l-gln-oh, carbobenzoxy-l-glutamine
PubChem CID 75855
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanoic acid
SMILES NC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 138°C
Color White
Quantity 25 g
Formula Weight 280.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.