Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-isoleucine 98.0+%, TCI America™
SDP

Catalog No. C135410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C135410G 10 g
C135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C135410G Supplier TCI America Supplier No. C135410G

Chemical Identifiers

CAS 3160-59-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00027064
InChI Key JSHXJPFZKBRLFU-JQWIXIFHSA-N
Synonym cbz-l-isoleucine, n-cbz-l-isoleucine, z-l-isoleucine, z-ile-oh, n-carbobenzyloxy-l-isoleucine, n-carbobenzoxy-l-isoleucine, cbz-l-ile-oh, n-benzyloxycarbonyl-l-isoleucine, l-isoleucine, n-phenylmethoxy carbonyl, 2s,3s-2-benzyloxy carbonyl amino-3-methylpentanoic acid
PubChem CID 2724772
IUPAC Name (2S,3S)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CCC(C)C(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 10 g
Formula Weight 265.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystals
Chemical Name or Material N-Carbobenzoxy-L-isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.