Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-methionine 98.0+%, TCI America™
SDP

Catalog No. C073725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C073725G 25 g
C07375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C073725G Supplier TCI America Supplier No. C073725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1152-62-1
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00065124
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Synonym z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
PubChem CID 1550857
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 25 g
Formula Weight 283.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.