Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-pyroglutamic Acid 98.0+%, TCI America™
SDP

Catalog No. C314325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C314325G 25 g
C31435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C314325G Supplier TCI America Supplier No. C314325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32159-21-0
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.25
MDL Number MFCD00037352
InChI Key VHSFUGXCSGOKJX-UHFFFAOYNA-N
Synonym z-pyr-oh, cbz-l-pyroglutamic acid, z-l-pyroglutamic acid, benzyloxycarbonyl-l-pyroglutamic acid, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, s-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid, z-pglu-oh
PubChem CID 637648
IUPAC Name 1-[(benzyloxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 141°C
Color White
Quantity 25 g
Formula Weight 263.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-pyroglutamic Acid
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.