Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxyglycine Methyl Ester 95.0+%, TCI America™
SDP

Catalog No. C283825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C283825G 25 g
C28385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C283825G Supplier TCI America Supplier No. C283825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1212-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00043631
InChI Key DZYBBBYFLOPVOL-UHFFFAOYSA-N
Synonym z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester
PubChem CID 71033
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}acetate
SMILES COC(=O)CNC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 151°C
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Carbobenzoxyglycine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.