Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxyhydroxylamine 97.0+%, TCI America™
SDP

Catalog No. C155725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C155725G 25 g
C15575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C155725G Supplier TCI America Supplier No. C155725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3426-71-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00010642
InChI Key PQBSPTAPCMSZAA-UHFFFAOYSA-N
Synonym benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
PubChem CID 18907
IUPAC Name benzyl N-hydroxycarbamate
SMILES ONC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 70°C
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxyhydroxylamine
RTECSNumber FB1600000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.