Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chlorosaccharin 96.0+%, TCI America™
SDP

Catalog No. C167425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C167425G 25 g
C16745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C167425G Supplier TCI America Supplier No. C167425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14070-51-0
Molecular Formula C7H4ClNO3S
Molecular Weight (g/mol) 217.623
MDL Number MFCD00274277
InChI Key VKWMGUNWDFIWNW-UHFFFAOYSA-N
Synonym n-chlorosaccharin, unii-g35nk0ctff, 2-chlorobenzo d isothiazol-3 2h-one 1,1-dioxide, g35nk0ctff, 1,2-benzisothiazol-3 2h-one, 2-chloro-, 1,1-dioxide, n-chlorosaccharine, ncsa, ncsac, n-chlorosaccharin mi, 2-chloro-1??,2-benzothiazole-1,1,3-trione
PubChem CID 4586058
IUPAC Name 2-chloro-1,1-dioxo-1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)N(S2(=O)=O)Cl

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 25 g
Formula Weight 217.62
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chlorosaccharin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.