Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cyclohexylmaleimide 98.0+%, TCI America™
SDP

Catalog No. C1044100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1044100G 100 g
C104425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1044100G Supplier TCI America Supplier No. C1044100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1631-25-0
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00043904
InChI Key BQTPKSBXMONSJI-UHFFFAOYSA-N
Synonym n-cyclohexylmaleimide, 1-cyclohexyl-1h-pyrrole-2,5-dione, maleimide, n-cyclohexyl, 1h-pyrrole-2,5-dione, 1-cyclohexyl, n-cyclohexyl maleimide, 1-cyclohexylazoline-2,5-dione, acmc-1btr9, 1h-pyrrole-2, 1-cyclohexyl, 1-cyclohexyl-pyrrole-2,5-dione
PubChem CID 74203
IUPAC Name 1-cyclohexyl-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1C1CCCCC1

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 132°C
Quantity 100 g
UN Number 2811
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Cyclohexylmaleimide
EINECSNumber (5)-5862
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.