Learn More
AdipoGen N-Desmethylthiamethoxam
SDP
Supplier:  AdipoGen CDXC0134M250
Encompass_Preferred
Chemical. CAS 171103-04-1. Formula C7H8ClN5O3S. MW 277.69. Synthetic. Neonicotinoid insecticide. Interacts with insect nicotinic acetylcholine receptors nAChRs, but is about 25 times less potent than thiamethoxam as an insecticide. Shown to be almost as active as the insecticide imidacloprid. Can be used as a reference compound.
Catalog No. 50-256-1463
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.