Learn More
SHOWA™ N-DEX™ 9005PF Nitrile Gloves
Catalog No. 11394115E Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-394-115E X-Large
11-394-115D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-394-115E Supplier SHOWA™ Supplier No. 9005PFXL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Single-use nitrile gloves offer tactile precision and high-capacity protection in a wide range of applications including automotive, chemical, municipalities, pharmaceutical and utilities.
  • Powder-free
  • Form-fitting
  • Ambidextrous
  • 100% Nitrile
  • Increased sensitivity
  • Rolled cuff, smooth grip
  • Ideal for agricultural, automotive, pharmaceutical, chemical, municipal applications
Order Info
N-DEX Original are packaged 100 gloves per dispenser, 20 dispensers per case (6005PF) or 100 gloves per polybag, 20 polybags per case (B6005PF). N-DEX Ultimate (9005PF) are packaged 50 pair per dispenser, 6 dispensers per case.
Specifications
Certifications/Compliance ASTM F 1671, ASTM D 6319, NFPA 1999, 21 CFR 170-199, FDA, ISO9002, GMP
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Glove Cuff Style Long
Model 9005PFXL
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.