Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecane, EMD-Millipore

Catalog No. M1096580005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096580005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096580005 Supplier MilliporeSigma Supplier No. 1.09658.0005

missing translation for 'validMainframeMsg'

Hydrocarbons C9 – C19

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 0.6 to 40 %(V)
Ignition Point 200°C
Vapor Pressure 0.4 hPa (20°C)
Viscosity <7mm2/sec., kinematic (40°C)
Density Density: 0.75g/cm3 (20°)
Assay Percent Range ≥99.0%
Linear Formula CH3(CH2)10CH3
Quantity 5 mL
Formula Weight 170.34
Chemical Name or Material n-Dodecane
Show More Show Less
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.