CAS RN 112-40-3
n-Dodecane, 99%, pure
CAS: 112-40-3 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
n-Dodecane, 99%
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
n-Dodecane, 99%, anhydrous, AcroSeal™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
N-dodecane, >99%, MP Biomedicals™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane, ≥99%, Reagent Grade, Solstice
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane 99.0+%, TCI America™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane 99.5+%, TCI America™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane, (n-Dodecane), Reagents
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-Dodecane, Dihexyl, Bihexyl IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane (Laboratory), Fisher Chemical
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 MDL Number: MFCD00008969 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane, 99% min., MilliporeSigma™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
MP Biomedicals, Inc n-Dodecane, MP Biomedicals™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N Synonym: n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661 PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
MilliporeSigma n-Dodecane, EMD-Millipore
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC
Dodecane, SPEX CertiPrep™
CAS: 112-40-3 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.34 InChI Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N PubChem CID: 8182 ChEBI: CHEBI:28817 IUPAC Name: dodecane SMILES: CCCCCCCCCCCC