Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyl 4-hydroxybenzoate, 98%, Thermo Scientific™

Catalog No. AAL0629206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0629206 5 g
AAL0629214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0629206 Supplier Thermo Scientific Chemicals Supplier No. L0629206

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2664-60-0
Molecular Formula C19H30O3
Molecular Weight (g/mol) 306.446
MDL Number MFCD00016473
InChI Key BAYSQTBAJQRACX-UHFFFAOYSA-N
Synonym dodecyl p-hydroxybenzoate, dodecylparaben, n-dodecyl 4-hydroxybenzoate, p-hydroxybenzoic acid dodecyl ester, benzoic acid, 4-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester, benzoic acid, p-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester german, 4-hydroxybenzoic acid dodecyl ester, laurylparaben
PubChem CID 17569
IUPAC Name dodecyl 4-hydroxybenzoate
SMILES CCCCCCCCCCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 54°C to 56°C
Quantity 5 g
Beilstein 2739355
Formula Weight 306.45
Percent Purity 98%
Chemical Name or Material n-Dodecyl 4-hydroxybenzoate
RTECSNumber DH2185600
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.