Learn More
AdipoGen n-Dodecyl-beta-D-maltoside
SDP
Supplier:  AdipoGen AGCC10004G025
Encompass_Preferred
Chemical. CAS 69227-93-6. Formula C24H46O11. MW 510.6. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.
Catalog No. 50-256-2717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.