Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethoxycarbonyl-5-nitro-o-toluidine 96.0+%, TCI America™
SDP

Catalog No. E034925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E034925G 25 g
E03495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E034925G Supplier TCI America Supplier No. E034925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16648-52-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00059827
InChI Key AYJFWTBDUMVOBQ-UHFFFAOYSA-N
Synonym N-Carbethoxy-5-nitro-o-toluidine, Ethyl 2-Methyl-5-nitrophenylcarbamate, N-(5-Nitro-2-methylphenyl)urethan
PubChem CID 601143
IUPAC Name ethyl N-(2-methyl-5-nitrophenyl)carbamate
SMILES CCOC(=O)NC1=C(C)C=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 224.22
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonyl-5-nitro-o-toluidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.