Learn More
Sigma Organic Chemistry N-Ethyl-2-naphthylamine | 100mg | MFCD00021600
Supplier: Sigma Organic Chemistry CDS011399100MG

N-Ethyl-2-naphthylamine , 100mg
About this Item:
MDL #: MFCD00021600
Molecular Weight: 171.24
UNSPSC Code: 12352200
Chemical Formula: C12H13N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.