Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Ethyl-N'-(3-dimethylaminopropyl)carbodiimide Hydrochloride, Condensing Reagent, Solid, 10 g

Catalog No. 502705454
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5454 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5454 Supplier Abcam Supplier No. AB14497910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble condensing reagent used as a carboxyl activating agent for peptide synthesis, protein crosslinking, and immunoconjugate preparation.

  • Molecular Weight: 191.7 g/mol
  • Form: Solid
  • Description: Water-soluble condensing reagent.
  • Applications: Generally utilized as a carboxyl activating agent for amide bonding with primary amines and reacts with phosphate groups.
  • Uses: Utilized in peptide synthesis, crosslinking proteins to nucleic acids, and preparation of immunoconjugates.
  • Storage Buffer: Constituents: 100% EDAC hydrochloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Carboxyl Groups, Primary Amines, Phosphate Groups, Proteins, Nucleic Acids
Kit Contents 10 g EDAC hydrochloride
Reactivity Carboxyl groups, primary amines, phosphate groups
Shipping Condition Ambient
Buffer 100% EDAC hydrochloride
Content And Storage Store at -20°C.
Form Solid
Formulation 100% EDAC hydrochloride
Quantity 10 g
For Use With (Application) Peptide synthesis, protein-nucleic acid crosslinking, immunoconjugate preparation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.