Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-N-(4-methylbenzyl)-2-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH6089703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6089703 1 g
AAH60897MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6089703 Supplier Thermo Scientific Chemicals Supplier No. H6089703

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421262-83-0
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
MDL Number MFCD22683152
InChI Key XPJKNBKPLZFMST-UHFFFAOYSA-N
Synonym n-ethyl-n-4-methylphenyl methyl-2-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-2-nitrobenzenesulfonamide
PubChem CID 97290229
IUPAC Name N-ethyl-N-[(4-methylphenyl)methyl]-2-nitrobenzenesulfonamide
SMILES CCN(CC1=CC=C(C)C=C1)S(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 334.39
Percent Purity 97%
Chemical Name or Material N-Ethyl-N-(4-methylbenzyl)-2-nitrobenzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.