Learn More
Medchemexpress LLC N-ethyl-N-oxido-dodecan-1-amine | 1643-20-5 | MFCD00002049 | 98.0% | 229.40 g/mol | C14H31NO | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1277851G
Encompass_Preferred
N-ethyl-N-oxido-dodecan-1-amine is an amine oxide surfactant used as a detergent, emulsifier, and wetting agent in research and industrial applications. It is a solid, white to off-white material with high purity and good solubility in common laboratory solvents, intended for research use.
  • High purity (98.0%).
  • Solid, white to off-white appearance.
  • Amine oxide surfactant useful as detergent and emulsifier.
  • Soluble in DMSO at approximately 100 mg/mL; may require sonication.
  • Stable when stored sealed and kept away from moisture.
  • Available in small laboratory packaging suitable for research use.
Catalog No. 50-002-83091