Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ N-Ferno™ 6480 Bottoms
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Specifications
Size | 2X-Large |
Color | Black |
For Use With (Application) | Road Construction, Heavy Construction,Building Construction, Iron/Steel Work, Oil/Gas Refining, Drilling/Mining, Freight/Baggage, Warehousing/Distribution, Landscaping/Grounds, Shakespeare In The Park |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title