Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ N-Ferno™ 6471 Thermal Bibs/Overalls

Catalog No. 19910198 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-198 2X-Large
19-910-194 Small
19-910-195 Medium
19-910-196 Large
19-910-197 X-Large
19-910-199 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-198 Supplier Ergodyne™ Supplier No. 41216
Add to Cart
Add to Cart
This item is not returnable. View return policy

Insulated bibs/overalls provide the ultimate in durability, weather protection, and quality outerwear

Features removable knee/shin pads and 1600D nylon reinforcement in heavy wear areas

Specifications

Color Black
Material Nylon (500D)
Size 2X-Large
For Use With (Application) Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.