Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ N-Ferno™ 6466 Thermal Jacket

Catalog No. 19910189 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-189 Medium
19-910-188 Small
19-910-190 Large
19-910-191 X-Large
19-910-192 2X-Large
19-910-193 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-189 Supplier Ergodyne™ Supplier No. 41113
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rugged 500D shell repels water and protects users from the wind and cold while remaining breathable for all day comfort.

For use with: Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics

Specifications

Size Medium
Material Nylon (500D)
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.