Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%

Catalog No. AAH6268003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6268003 1 g
AAH62680MD 250 mg
AAH6268006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6268003 Supplier Thermo Scientific Chemicals Supplier No. H6268003
Add to Cart

CAS: 134098-70-7 | C23H25NO6 | 411.454 g/mol

N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134098-70-7
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.454
MDL Number MFCD00237011
InChI Key VZXQYACYLGRQJU-LJQANCHMSA-N
Synonym fmoc-d-asp-otbu, fmoc-d-aspartic acid 1-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid alpha-t-butyl ester, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp otbu, ambotzfaa1311, fmoc-d-aspartic acid-otbu, fmoc-d-asp-tert-butoxide, fmoc-d-aspartic acid alpha-t-butyl ester
PubChem CID 7019703
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 1 g
Solubility Information Soluble in DMF.
Formula Weight 411.45
Percent Purity 95%
Chemical Name or Material N-Fmoc-D-aspartic acid 1-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.