Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-glutamic acid 1-tert-butyl ester, 98%

Catalog No. AAH6214603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6214603 1 g
AAH6214606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6214603 Supplier Thermo Scientific Chemicals Supplier No. H6214603
Add to Cart

CAS: 84793-07-7 | C24H27NO6 | 425.481 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84793-07-7
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.481
MDL Number MFCD00065648
InChI Key GOPWHXPXSPIIQZ-FQEVSTJZSA-N
Synonym fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b
PubChem CID 7017912
IUPAC Name (4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 1 g
Formula Weight 425.47
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-glutamic acid 1-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.