Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-glutamic acid, 95%

Catalog No. AAH6293614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6293614 25 g
AAH6293606 5 g
AAH6293622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6293614 Supplier Thermo Scientific Chemicals Supplier No. H6293614
Add to Cart

CAS: 121343-82-6 | C20H19NO6 | 369.373 g/mol

N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121343-82-6
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
MDL Number MFCD00237657
InChI Key QEPWHIXHJNNGLU-KRWDZBQOSA-N
Synonym fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid
PubChem CID 7019018
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Quantity 25 g
Beilstein 3633624
Solubility Information Slightly soluble in water.
Formula Weight 369.37
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-glutamic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.